Thermo Scientific Chemicals

Diethylbenzene, Mixture of o-, m- and p-isomers, Sum Min. 95%, Thermo Scientific Chemicals

Catalog number: 114160010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Diethylbenzene, Mixture of o-, m- and p-isomers, Sum Min. 95%, Thermo Scientific Chemicals

Catalog number: 114160010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 114160010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
25340-17-4
IUPAC Name
1,2-diethylbenzene
Molecular Formula
C10H14
InChI Key
KVNYFPKFSJIPBJ-UHFFFAOYSA-N
SMILES
CCC1=CC=CC=C1CC
Appearance (Form)
Liquid
Refractive index
1.4940 to 1.4960 (20°C, 589 nm)
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to light yellow
GC
>=95.0 % (sum of o-, m-, p-isomers)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text