Diethylbenzene, Mixture of o-, m- and p-isomers, Sum Min. 95%, Thermo Scientific Chemicals
Diethylbenzene, Mixture of o-, m- and p-isomers, Sum Min. 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethylbenzene, Mixture of o-, m- and p-isomers, Sum Min. 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Glass bottle
Catalog number 114160025
Price (USD)/ Each
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1,2-diethylbenzene
Molecular FormulaC10H14
InChI KeyKVNYFPKFSJIPBJ-UHFFFAOYSA-N
SMILESCCC1=CC=CC=C1CC
Molecular Weight (g/mol)134.22
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4940 to 1.4960 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
GC>=95.0 % (sum of o-, m-, p-isomers)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only