Diethyl benzylphosphonate, 99%
Diethyl benzylphosphonate, 99%
Diethyl benzylphosphonate, 99%
Diethyl benzylphosphonate, 99%
Thermo Scientific Chemicals

Diethyl benzylphosphonate, 99%

CAS: 1080-32-6 | C11H17O3P | 228.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 114170250
Price (USD)
74.65
Online Exclusive
83.20
Save 8.55 (10%)
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
74.65
Online Exclusive
83.20
Save 8.55 (10%)
Each
Add to cart
Chemical Identifiers
CAS1080-32-6
IUPAC Namediethyl benzylphosphonate
Molecular FormulaC11H17O3P
InChI KeyAIPRAPZUGUTQKX-UHFFFAOYSA-N
SMILESCCOP(=O)(CC1=CC=CC=C1)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4950 to 1.4980 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only

Share catalog number, name or link