Thermo Scientific Chemicals

Diethyl bromomalonate, 90+%, Thermo Scientific Chemicals

Catalog number: 114190250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl bromomalonate, 90+%, Thermo Scientific Chemicals

Catalog number: 114190250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 114190250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
685-87-0
IUPAC Name
1,3-diethyl 2-bromopropanedioate
Molecular Formula
C7H11BrO4
InChI Key
FNJVDWXUKLTFFL-UHFFFAOYSA-N
SMILES
CCOC(=O)C(Br)C(=O)OCC
Appearance (Form)
Liquid
Refractive index
1.4510 to 1.4560 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
GC
>=90.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text