Diethyl bromomalonate, 90+%, Thermo Scientific Chemicals
Diethyl bromomalonate, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl bromomalonate, 90+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 114195000
Price (USD)/ Each
509.00
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
509.00
Add to cart
Diethyl bromomalonate, 90+%, Thermo Scientific Chemicals
Catalog number114195000
Price (USD)/ Each
509.00
-
Add to cart
Chemical Identifiers
CAS685-87-0
IUPAC Name1,3-diethyl 2-bromopropanedioate
Molecular FormulaC7H11BrO4
InChI KeyFNJVDWXUKLTFFL-UHFFFAOYSA-N
SMILESCCOC(=O)C(Br)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=90.0 %
Appearance (Form)Liquid
Refractive index1.4510 to 1.4560 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only