Diethyl carbonate, 99%, Thermo Scientific Chemicals
Diethyl carbonate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl carbonate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
100 mL
500 mL
1 L
2.5 L
Packaging:
View all
Plastic drum
Catalog number 114220100
Price (USD)/ Each
514.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
605.00 
Save 90.35 (15%)
-
Add to cart
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
514.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
605.00 
Save 90.35 (15%)
Add to cart
Diethyl carbonate, 99%, Thermo Scientific Chemicals
Catalog number114220100
Price (USD)/ Each
514.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
605.00 
Save 90.35 (15%)
-
Add to cart
Chemical Identifiers
CAS105-58-8
IUPAC Namediethyl carbonate
Molecular FormulaC5H10O3
InChI KeyOIFBSDVPJOWBCH-UHFFFAOYSA-N
SMILESCCOC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Refractive index1.3830 to 1.3860 (20°C, 589 nm)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Alkylation reagent
RUO – Research Use Only