Thermo Scientific Chemicals

Diethyl cyanomethylphosphonate, 96%, Thermo Scientific Chemicals

Catalog number: 114272500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl cyanomethylphosphonate, 96%, Thermo Scientific Chemicals

Catalog number: 114272500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 114272500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2537-48-6
IUPAC Name
diethyl (cyanomethyl)phosphonate
Molecular Formula
C6H12NO3P
InChI Key
KWMBADTWRIGGGG-UHFFFAOYSA-N
SMILES
CCOP(=O)(CC#N)OCC
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.4312 to 1.4332 (20°C, 589 nm)
Infrared spectrum
Conforms
GC
>=95 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text