Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 114290050
Price (USD)/ Each
71.65
Online exclusive
83.80 
Save 12.15 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
71.65
Online exclusive
83.80 
Save 12.15 (14%)
Add to cart
Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals
Catalog number114290050
Price (USD)/ Each
71.65
Online exclusive
83.80 
Save 12.15 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Namesilver(1+) (diethylcarbamothioyl)sulfanide
Molecular FormulaC5H10AgNS2
InChI KeyNSVHDIYWJVLAGH-UHFFFAOYSA-M
SMILES[Ag+].CCN(CC)C([S-])=S
Molecular Weight (g/mol)256.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to green to khaki
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NH4SCN98.5 to 101.5 %
Solubility(0.5% in pyridine) clear
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only