Diethylenetriamine, 98+%, Thermo Scientific Chemicals
Diethylenetriamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethylenetriamine, 98+%, Thermo Scientific Chemicals

Diethylenetriamine, 98+%, C4H13N3, CAS Number-111-40-0, 1,4,7-triazaheptane, 2,2'-diaminodiethylamine, diethylenetriamine, 2,2'-iminodiethylamine, bis 2-aminoethyl amine, barsamide 115, ancamine deta, n,n-bis 2-aminoethyl amine, epicure t, diethylene triamine, 2.5L, 98% min. (GC), 04,255
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
10 L
Packaging:
View all
Glass bottle
Catalog number 114310025
Price (USD)/ Each
126.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
149.00 
Save 22.35 (15%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
126.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
149.00 
Save 22.35 (15%)
Add to cart
Diethylenetriamine, 98+%, Thermo Scientific Chemicals
Catalog number114310025
Price (USD)/ Each
126.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
149.00 
Save 22.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namebis(2-aminoethyl)amine
Molecular FormulaC4H13N3
InChI KeyRPNUMPOLZDHAAY-UHFFFAOYSA-N
SMILESNCCNCCN
Molecular Weight (g/mol)103.17
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<50 APHA
Infrared spectrumConforms
Refractive index1.4820 to 1.4850 (20°C, 589 nm)
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only