Diethylenetriaminepentaacetic acid, 98+%
Diethylenetriaminepentaacetic acid, 98+%
Diethylenetriaminepentaacetic acid, 98+%
Diethylenetriaminepentaacetic acid, 98+%
Thermo Scientific Chemicals

Diethylenetriaminepentaacetic acid, 98+%

CAS: 67-43-6 | C14H23N3O10 | 393.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 114320050
Price (USD)
314.65
Special offer
Online exclusive
370.00
Save 55.35 (15%)
Each
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
314.65
Special offer
Online exclusive
370.00
Save 55.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS67-43-6
IUPAC Name2-[bis({2-[bis(carboxymethyl)amino]ethyl})amino]acetic acid
Molecular FormulaC14H23N3O10
InChI KeyQPCDCPDFJACHGM-UHFFFAOYSA-N
SMILESOC(=O)CN(CCN(CC(O)=O)CC(O)=O)CCN(CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white
Titration excess NaOH>=98.0 %
Appearance (Form)Crystalline powder
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470235Certificate of AnalysisMay 17, 2025114322500, 114320010, 114320050
A0465643Certificate of AnalysisAug 26, 2024114322500, 114320010, 114320050
A0398555Certificate of AnalysisDec 15, 2022114322500, 114320010, 114320050
A0401517Certificate of AnalysisDec 15, 2022114322500, 114320010, 114320050
A0401516Certificate of AnalysisDec 15, 2022114322500, 114320010, 114320050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection