Diethylenetriaminepentaacetic acid, 98+%, Thermo Scientific Chemicals
Diethylenetriaminepentaacetic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethylenetriaminepentaacetic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 114322500
Price (USD)/ Each
36.65
Online exclusive
40.80 
Save 4.15 (10%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
36.65
Online exclusive
40.80 
Save 4.15 (10%)
Add to cart
Diethylenetriaminepentaacetic acid, 98+%, Thermo Scientific Chemicals
Catalog number114322500
Price (USD)/ Each
36.65
Online exclusive
40.80 
Save 4.15 (10%)
-
Add to cart
Chemical Identifiers
CAS67-43-6
IUPAC Name2-[bis({2-[bis(carboxymethyl)amino]ethyl})amino]acetic acid
Molecular FormulaC14H23N3O10
InChI KeyQPCDCPDFJACHGM-UHFFFAOYSA-N
SMILESOC(=O)CN(CCN(CC(O)=O)CC(O)=O)CCN(CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white
Titration excess NaOH>=98.0 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only