Thermo Scientific Chemicals

Diethyl ethylmalonate, 99%, Thermo Scientific Chemicals

Catalog number: 114361000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl ethylmalonate, 99%, Thermo Scientific Chemicals

Catalog number: 114361000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 114361000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
133-13-1
IUPAC Name
1,3-diethyl 2-ethylpropanedioate
Molecular Formula
C9H16O4
InChI Key
VQAZCUCWHIIFGE-UHFFFAOYSA-N
SMILES
CCOC(=O)C(CC)C(=O)OCC
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.4150 to 1.4170 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text