Thermo Scientific Chemicals

Diethyl fumarate, 98%, Thermo Scientific Chemicals

Catalog number: 114380010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl fumarate, 98%, Thermo Scientific Chemicals

Catalog number: 114380010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 114380010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
623-91-6
IUPAC Name
1,4-diethyl (2E)-but-2-enedioate
Molecular Formula
C8H12O4
InChI Key
IEPRKVQEAMIZSS-AATRIKPKSA-N
SMILES
CCOC(=O)\C=C\C(=O)OCC
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
GC
>=97.5 %
Infrared spectrum
Conforms
Refractive index
1.4396 to 1.4416 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text