Thermo Scientific Chemicals

N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals

Catalog number: 114421000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals

Catalog number: 114421000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 114421000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3710-84-7
IUPAC Name
N,N-diethylhydroxylamine
Molecular Formula
C4H11NO
InChI Key
FVCOIAYSJZGECG-UHFFFAOYSA-N
SMILES
CCN(O)CC
Appearance (Form)
Clear liquid
Color scale
=<100 APHA
GC
>=96.0 %
Refractive index
1.4190 to 1.4220 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text