Diethyl maleate, 97%, Thermo Scientific Chemicals
Diethyl maleate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl maleate, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
250 g
Packaging:
Glass bottle
Catalog number 114442500
Price (USD)/ Each
32.65
Online exclusive
36.40 
Save 3.75 (10%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
32.65
Online exclusive
36.40 
Save 3.75 (10%)
Add to cart
Diethyl maleate, 97%, Thermo Scientific Chemicals
Catalog number114442500
Price (USD)/ Each
32.65
Online exclusive
36.40 
Save 3.75 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Name1,4-diethyl (2Z)-but-2-enedioate
Molecular FormulaC8H12O4
InChI KeyIEPRKVQEAMIZSS-WAYWQWQTSA-N
SMILESCCOC(=O)\C=C/C(=O)OCC
Molecular Weight (g/mol)172.18
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4380 to 1.4420 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only