Diethyl malonate, 99+%, Thermo Scientific Chemicals
Diethyl malonate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl malonate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
2.5 kg
Packaging:
View all
Plastic drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 114450100
Price (USD)/ Each
331.00
-
Add to cart
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
331.00
Add to cart
Diethyl malonate, 99+%, Thermo Scientific Chemicals
Catalog number114450100
Price (USD)/ Each
331.00
-
Add to cart
Chemical Identifiers
CAS105-53-3
IUPAC Name1,3-diethyl propanedioate
Molecular FormulaC7H12O4
InChI KeyIYXGSMUGOJNHAZ-UHFFFAOYSA-N
SMILESCCOC(=O)CC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Density(20°C) 1.052 to 1.058 g/ml
Appearance (Form)Liquid
Refractive index1.4120 to 1.4160 (20°C, 589 nm)
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only