2,4-Difluoroaniline, 99%
2,4-Difluoroaniline, 99%
2,4-Difluoroaniline, 99%
2,4-Difluoroaniline, 99%
Thermo Scientific Chemicals

2,4-Difluoroaniline, 99%

CAS: 367-25-9 | C6H5F2N | 129.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
25 mL
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 114590100
Price (USD)
-
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS367-25-9
IUPAC Name2,4-difluoroaniline
Molecular FormulaC6H5F2N
InChI KeyCEPCPXLLFXPZGW-UHFFFAOYSA-N
SMILESNC1=CC=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
AppearanceMay darken on storage
Refractive index1.5055 to 1.5085 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link