Thermo Scientific Chemicals

2,4-Difluoroaniline, 99%, Thermo Scientific Chemicals

Catalog number: 114590250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

2,4-Difluoroaniline, 99%, Thermo Scientific Chemicals

Catalog number: 114590250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 114590250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
367-25-9
IUPAC Name
2,4-difluoroaniline
Molecular Formula
C6H5F2N
InChI Key
CEPCPXLLFXPZGW-UHFFFAOYSA-N
SMILES
NC1=CC=C(F)C=C1F
Appearance (Color)
Clear colorless to yellow to brown
Appearance
May darken on storage
Refractive index
1.5055 to 1.5085 (20°C, 589 nm)
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text