Thermo Scientific Chemicals

sym-Diformylhydrazine, 97%, Thermo Scientific Chemicals

Catalog number: 114640250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

sym-Diformylhydrazine, 97%, Thermo Scientific Chemicals

Catalog number: 114640250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 114640250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
628-36-4
IUPAC Name
N'-formylformohydrazide
Molecular Formula
C2H4N2O2
InChI Key
POVXOWVFLAAVBH-UHFFFAOYSA-N
SMILES
O=CNNC=O
Appearance (Form)
Crystalline powder
TLC
>=96.0 %
Infrared spectrum
Conforms
Appearance (Color)
White to off-white
Melting point
155°C to 160°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text