Diglycolic anhydride, 97%, Thermo Scientific Chemicals
Diglycolic anhydride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diglycolic anhydride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 114670250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1,4-dioxane-2,6-dione
Molecular FormulaC4H4O4
InChI KeyPIYNUZCGMLCXKJ-UHFFFAOYSA-N
SMILESO=C1COCC(=O)O1
Molecular Weight (g/mol)116.07
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Melting point90°C to 95°C
Appearance (Form)Crystalline powder or crystals or needles
Infrared spectrumConforms
Titration excess NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only