Thermo Scientific Chemicals

2,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 114870050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

2,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 114870050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 114870050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
89-86-1
IUPAC Name
2,4-dihydroxybenzoic acid
Molecular Formula
C7H6O4
InChI Key
UIAFKZKHHVMJGS-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(O)C=C1O
Appearance (Form)
Crystalline powder
HPLC
>=96.0 %
Appearance (Color)
Cream to light beige to pink
Titration with NaOH
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text