2,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals
2,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number 114875000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS89-86-1
IUPAC Name2,4-dihydroxybenzoic acid
Molecular FormulaC7H6O4
InChI KeyUIAFKZKHHVMJGS-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(O)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Appearance (Color)Cream to light beige to pink
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only