Thermo Scientific Chemicals

3,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 114890250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 114890250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 114890250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-50-3
IUPAC Name
3,4-dihydroxybenzoic acid
Molecular Formula
C7H6O4
InChI Key
YQUVCSBJEUQKSH-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(O)C(O)=C1
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
198°C to 202°C
Titration with NaOH
>=96.0 %
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text