Thermo Scientific Chemicals

3,4-Dihydroxycinnamic acid, 99+%, predominantly trans isomer, Thermo Scientific Chemicals

Catalog number: 114930050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3,4-Dihydroxycinnamic acid, 99+%, predominantly trans isomer, Thermo Scientific Chemicals

Catalog number: 114930050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 114930050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
331-39-5
IUPAC Name
(2Z)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
Molecular Formula
C9H8O4
InChI Key
QAIPRVGONGVQAS-RQOWECAXSA-N
SMILES
OC(=O)\C=C/C1=CC=C(O)C(O)=C1
Appearance (Form)
Powder
Titration with NaOH
>=99.0 %
Appearance (Color)
Yellow to yellow-green
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text