3,4-Dihydroxycinnamic acid, 99+%, predominantly trans isomer, Thermo Scientific Chemicals
3,4-Dihydroxycinnamic acid, 99+%, predominantly trans isomer, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Dihydroxycinnamic acid, 99+%, predominantly trans isomer, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 114930250
Price (USD)/ Each
135.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
159.00 
Save 23.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
135.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
159.00 
Save 23.35 (15%)
Add to cart
3,4-Dihydroxycinnamic acid, 99+%, predominantly trans isomer, Thermo Scientific Chemicals
Catalog number114930250
Price (USD)/ Each
135.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
159.00 
Save 23.35 (15%)
-
Add to cart
Chemical Identifiers
CAS331-39-5
IUPAC Name(2Z)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
Molecular FormulaC9H8O4
InChI KeyQAIPRVGONGVQAS-RQOWECAXSA-N
SMILESOC(=O)\C=C/C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with NaOH>=99.0 %
Appearance (Color)Yellow to yellow-green
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only