Thermo Scientific Chemicals

1,5-Dihydroxynaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: 115005000
500 g, Each
Thermo Scientific Chemicals

1,5-Dihydroxynaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: 115005000
500 g, Each
Quantity
Catalog number: 115005000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
83-56-7
IUPAC Name
naphthalene-1,5-diol
Molecular Formula
C10H8O2
InChI Key
BOKGTLAJQHTOKE-UHFFFAOYSA-N
SMILES
OC1=CC=CC2=C(O)C=CC=C12
Appearance (Color)
Grey-brown to brown
HPLC
>=96.0 %
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text