Thermo Scientific Chemicals

7-(2,3-Dihydroxypropyl)theophylline, 99%, Thermo Scientific Chemicals

Catalog number: 115055000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

7-(2,3-Dihydroxypropyl)theophylline, 99%, Thermo Scientific Chemicals

Catalog number: 115055000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 115055000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
479-18-5
IUPAC Name
7-(2,3-dihydroxypropyl)-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular Formula
C10H14N4O4
InChI Key
KSCFJBIXMNOVSH-UHFFFAOYNA-N
SMILES
CN1C2=C(N(CC(O)CO)C=N2)C(=O)N(C)C1=O
Melting point
160°C to 165°C
Titration with HClO4
>=98.5 % (On dried substance)
Loss on drying
=<0.5 % (1 g, 105°C)
Appearance (Color)
White
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text