4,6-Dihydroxypyrimidine, 98%, Thermo Scientific Chemicals
4,6-Dihydroxypyrimidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,6-Dihydroxypyrimidine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number 115085000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name6-hydroxy-3,4-dihydropyrimidin-4-one
Molecular FormulaC4H4N2O2
InChI KeyDUFGYCAXVIUXIP-UHFFFAOYSA-N
SMILESOC1=CC(=O)NC=N1
Molecular Weight (g/mol)112.09
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Infrared spectrumConforms
Titration excess NaOH>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only