1,2-Diiodoethane, 99%, Thermo Scientific Chemicals
1,2-Diiodoethane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Diiodoethane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 115141000
Price (USD)/ Each
154.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 27.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
154.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 27.35 (15%)
Add to cart
1,2-Diiodoethane, 99%, Thermo Scientific Chemicals
Catalog number115141000
Price (USD)/ Each
154.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 27.35 (15%)
-
Add to cart
Chemical Identifiers
CAS624-73-7
IUPAC Name1,2-diiodoethane
Molecular FormulaC2H4I2
InChI KeyGBBZLMLLFVFKJM-UHFFFAOYSA-N
SMILESICCI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point80°C to 83°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only