1,6-Diisocyanatohexane, 99+%
1,6-Diisocyanatohexane, 99+%
1,6-Diisocyanatohexane, 99+%
1,6-Diisocyanatohexane, 99+%
Thermo Scientific Chemicals

1,6-Diisocyanatohexane, 99+%

CAS: 822-06-0 | C8H12N2O2 | 168.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 115190250
Price (USD)
51.10
Each
In stock
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
51.10
Each
Add to cart
Chemical Identifiers
CAS822-06-0
IUPAC Name1,6-diisocyanatohexane
Molecular FormulaC8H12N2O2
InChI KeyRRAMGCGOFNQTLD-UHFFFAOYSA-N
SMILESO=C=NCCCCCCN=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.4520 to 1.4540 (20°C, 589 nm)
Infrared spectrumConforms
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection