Thermo Scientific Chemicals

N,N'-Diisopropylcarbodiimide, 99%, Thermo Scientific Chemicals

Catalog number: 115210250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

N,N'-Diisopropylcarbodiimide, 99%, Thermo Scientific Chemicals

Catalog number: 115210250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 115210250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
693-13-0
IUPAC Name
(propan-2-yl)({[(propan-2-yl)imino]methylidene})amine
Molecular Formula
C7H14N2
InChI Key
BDNKZNFMNDZQMI-UHFFFAOYSA-N
SMILES
CC(C)N=C=NC(C)C
GC
>=98.5 %
Appearance (Form)
Liquid
Refractive index
1.4320 to 1.4350 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text