N,N-Diisopropylethylamine, 99+%
N,N-Diisopropylethylamine, 99+%
N,N-Diisopropylethylamine, 99+%
N,N-Diisopropylethylamine, 99+%
Thermo Scientific Chemicals

N,N-Diisopropylethylamine, 99+%

CAS: 7087-68-5 | C8H19N | 129.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
100 g
10 kg
Packaging:
View all
Glass bottle
Catalog number 115220250
Price (USD)
51.80
Each
In stock
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
51.80
Each
Add to cart
Chemical Identifiers
CAS7087-68-5
IUPAC Nameethylbis(propan-2-yl)amine
Molecular FormulaC8H19N
InChI KeyJGFZNNIVVJXRND-UHFFFAOYSA-N
SMILESCCN(C(C)C)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4120 to 1.4150 (20°C, 589 nm)
Diisopropylamine=<0.30 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473702Certificate of AnalysisJun 25, 2025115220101, 115221000, 115220250, 115225000, 115220026
A0463104Certificate of AnalysisMay 26, 2025115220101, 115221000, 115220250, 115225000, 115220026
A0467840Certificate of AnalysisJan 31, 2025115220101, 115221000, 115220250, 115225000, 115220026
A0468860Certificate of AnalysisJan 31, 2025115220101, 115221000, 115220250, 115225000, 115220026
A0469342Certificate of AnalysisJan 30, 2025115220101, 115221000, 115220250, 115225000, 115220026
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection