Interferon alpha-21 Recombinant Protein
Interferon alpha-21 Recombinant Protein

Interferon alpha-21 Recombinant Protein

Human Leukocyte Interferon Recombinant Protein for Western Blot, IP, ELISARead more
Have Questions?
Catalog number 115261000
Price (USD)/ Each
-
Request bulk or custom format
Specifications
Chemical Name or MaterialDiisopropyl phosphite
Name Note98%
CAS1809-20-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
View more
Human Leukocyte Interferon Recombinant Protein for Western Blot, IP, ELISA