Search Thermo Fisher Scientific
The ELEMENT Series HR-ICP-MS system is a potent multi-elemental detector for transient signals, such as those from laser ablation analysis, making this an ideal solids analysis solution.
Type | Laser Ablation Adaption Kit |
For Use With (Equipment) | ELEMENT Series HR-ICP-MS System |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
1152810 | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
This useful Laser Ablation Adaptation Kit includes an additional He mass flow controller, adaptors for coupling the laser to the sample inlet system and a communication cable to trigger the laser ablation system with the mass spectrometer.
The ELEMENT Series HR-ICP-MS system is compatible with laser ablation systems from, amongst others, Photon Machines, Thermo Scientific™ CETAC™ and esi™ New Wave Research.