Save up to 49% with protein expression workflow promo

Save now

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • Creatine Kinase BB Antibodies

            NeoBiotechnologies

            Creatine Phosphokinase-BB (CK-BB) Monoclonal Antibody (CKBB/6871)

            View all (54) Creatine Kinase BB antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Catalog # 1152-MSM11-P1ABX
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (KRW)
            903,000
            100 µg
             
            Check your price
            Request bulk or custom format
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite Creatine Phosphokinase-BB (CK-BB) Monoclonal Antibody (CKBB/6871)

            • Antibody Testing Data (1)
            Application
            Creatine Phosphokinase-BB (CK-BB) Antibody in SDS-PAGE (SDS-PAGE)
            Group 53 Created with Sketch.

            FIGURE: 1 / 1

            Creatine Phosphokinase-BB (CK-BB) Antibody (1152-MSM11-P1ABX) in SDS-PAGE

            SDS-PAGE analysis Purified CKBB Mouse Monoclonal Antibody (CKBB/6871). Confirmation of Purity and Integrity of Antibody.
            View Product

            Creatine Phosphokinase-BB (CK-BB) Antibody in SDS-PAGE (SDS-PAGE)

            Product Details

            1152-MSM11-P1ABX

            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Mouse / IgG2b, kappa

            Class

            Monoclonal

            Type

            Antibody

            Clone

            CKBB/6871

            Immunogen

            Recombinant human full-length CKBprotein
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Protein A/G

            Storage buffer

            PBS, pH 7.4

            Contains

            no preservative

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Target Information

            The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: B-CK; Brain creatine kinase; CPK-B; Creatine kinase B chain; Creatine kinase B-type; creatine kinase brain; creatine kinase brain-type; creatine kinase, brain; creatine kinase-B; Creatine phosphokinase B-type; epididymis luminal protein 211; epididymis secretory protein Li 29

            View more View less

            Gene Aliases: B-CK; BCK; CKB; CKBB; HEL-211; HEL-S-29

            View more View less

            UniProt ID: (Human) P12277

            View more View less

            Entrez Gene ID: (Human) 1152

            View more View less

            Function(s)

            creatine kinase activity protein binding ATP binding ubiquitin protein ligase binding amino acid kinase

            Process(es)

            creatine metabolic process phosphorylation substantia nigra development cellular chloride ion homeostasis
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            • Fair Trade
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            korea | 대한민국 flag icon
            Korea | 대한민국

            Customer Help Desk | working day 09:00~18:00
            1661-9555   |   Live Chat   |   카카오톡 상담

             

            Service Call Center | working day 09:00~18:00
            1661-5055   |   Live Chat

            Customer Help Desk | working day 09:00~18:00
            1661-9555   |   Live Chat   |   카카오톡 상담

             

            Service Call Center | working day 09:00~18:00
            1661-5055   |   Live Chat

            Customer Help Desk | working day 09:00~18:00
            1661-9555   |   Live Chat   |   카카오톡 상담

             

            Service Call Center | working day 09:00~18:00
            1661-5055   |   Live Chat

            Thermo Fisher Scientific Korea Ltd.
            Representative : Soojin Seok
            Company Registration No. : 117-81-46910

             

            Thermo Fisher Scientific Solutions LLC
            Representative : Soojin Seok
            Company Registration No. : 114-86-04783

             

            Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-5zdmk:80/100.66.79.0:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0