Invitrogen™

CELLection™ Pan Mouse IgG Kit

Catalog number: 11531D
Have Questions?
Invitrogen™

CELLection™ Pan Mouse IgG Kit

Catalog number: 11531D
Have Questions?
Catalog Number
11531D
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
11531D5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Use this kit with any mouse IgG for the positive isolation of a cell population from all species except mouse. Antibodies are attached to the surface of the CELLection™ Dynabeads™ via a DNA linker; this linker provides a cleavable site to release and remove the beads from the cells after isolation. Bead-free cells are viable and can be used in any downstream application.

This Kit Contains: CELLection™ Dynabeads™ coated with a human anti-mouse IgG monoclonal, and DNase Releasing Buffer

Applications:

• Positively isolate and detach any cell from any species (except mouse) using your own mouse IgG
• Compatible with flow cytometry, cell culture and all cell based assays
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Antibody Fragment
Whole Antibody
Cell Type
All cells from all species, except mouse
Product Type
Antibody Coated Bead
Dye Type
CELLection™ Pan
Host Species
Mouse
Isolation Technology
Positive Isolation
No. of Cells
Processes ∼2 x 109 cells total
Output Viability
>95%
Product Line
CELLection™, Dynabeads™
Purity or Quality Grade
Research Grade
Reactivity
All species, except mouse
Sample Type
PBMC, Tissue Digests, Blood
Shipping Condition
Room Temperature
Starting Material Cell No.
1 x 107 PBMCs per isolation
Target Species
All species, except mouse

Contents & Storage

Store in refrigerator (2–8°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text