Invitrogen™

CELLection™ Biotin Binder Kit

Catalog number: 11533D
Have Questions?
Invitrogen™

CELLection™ Biotin Binder Kit

Catalog number: 11533D
Have Questions?
Catalog Number
11533D
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
11533D5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Use this kit with any biotinylated antibody for the positive isolation of a cell population including mouse cells. Biotinylated antibodies are attached to CELLection™ Dynabeads™ via a DNA linker; this linker provides a cleavable site to remove the beads from the cells after isolation. Detached cells are viable and can be used in any downstream application.

This Product Contains: CELLection™ Dynabeads™ coated with recombinant streptavidin and DNase Releasing Buffer

Applications:

• Positively isolate and detach any cell from any species using your own biotinylated antibody
• Compatible with flow cytometry
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
All cells from all species
Product Type
Biotin Binder
Isolation Technology
Positive Isolation
No. of Cells
Processes ∼2 x 109 cells total
Output Viability
>95%
Product Line
CELLection™, Dynabeads™
Purity or Quality Grade
Research Grade
Reactivity
All species
Sample Type
PBMC, Tissue Digests, Blood
Shipping Condition
Room Temperature
Starting Material Cell No.
1 x 107 PBMCs per isolation
Target Species
All species

Contents & Storage

Store in refrigerator (2–8°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text