Thermo Scientific Chemicals

3,4-Dimethoxycinnamic acid, 99%, predominantly trans isomer, Thermo Scientific Chemicals

Catalog number: 115511000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

3,4-Dimethoxycinnamic acid, 99%, predominantly trans isomer, Thermo Scientific Chemicals

Catalog number: 115511000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 115511000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2316-26-9
IUPAC Name
(2E)-3-(3,4-dimethoxyphenyl)prop-2-enoic acid
Molecular Formula
C11H12O4
InChI Key
HJBWJAPEBGSQPR-GQCTYLIASA-N
SMILES
COC1=CC=C(C=CC(O)=O)C=C1OC
Melting point
181°C to 184°C
Appearance (Color)
Light yellow to light beige
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with NaOH
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text