2,5-Dimethoxy-2,5-dihydrofuran, 97%, mixture of cis and trans, Thermo Scientific Chemicals
2,5-Dimethoxy-2,5-dihydrofuran, 97%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,5-Dimethoxy-2,5-dihydrofuran, 97%, mixture of cis and trans, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 g
Packaging:
Glass bottle
Catalog number 115531000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2,5-dimethoxy-2,5-dihydrofuran
Molecular FormulaC6H10O3
InChI KeyWXFWXFIWDGJRSC-UHFFFAOYNA-N
SMILESCOC1OC(OC)C=C1
Molecular Weight (g/mol)130.14
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4330 to 1.4350 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only