Thermo Scientific Chemicals

3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer, Thermo Scientific Chemicals

Catalog number: 115550250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer, Thermo Scientific Chemicals

Catalog number: 115550250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 115550250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
530-59-6
IUPAC Name
(2E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
Molecular Formula
C11H12O5
InChI Key
PCMORTLOPMLEFB-ONEGZZNKSA-N
SMILES
COC1=CC(C=CC(O)=O)=CC(OC)=C1O
Appearance (Color)
Yellow to light beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text