Thermo Scientific Chemicals

(3,4-Dimethoxyphenyl)acetic acid, 99%, Thermo Scientific Chemicals

Catalog number: 115611000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

(3,4-Dimethoxyphenyl)acetic acid, 99%, Thermo Scientific Chemicals

Catalog number: 115611000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 115611000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
93-40-3
IUPAC Name
2-(3,4-dimethoxyphenyl)acetic acid
Molecular Formula
C10H12O4
InChI Key
WUAXWQRULBZETB-UHFFFAOYSA-N
SMILES
COC1=CC=C(CC(O)=O)C=C1OC
Water
=<1 % (K.F.)
Appearance (Color)
White to beige
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with TBAH
>=98.5 % (On dry substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text