2,5-Dimethoxytetrahydrofuran, 99%, mixture of cis- and trans isomers, Thermo Scientific Chemicals
2,5-Dimethoxytetrahydrofuran, 99%, mixture of cis- and trans isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,5-Dimethoxytetrahydrofuran, 99%, mixture of cis- and trans isomers, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number 115675000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS696-59-3
IUPAC Name2,5-dimethoxyoxolane
Molecular FormulaC6H12O3
InChI KeyGFISDBXSWQMOND-UHFFFAOYNA-N
SMILESCOC1CCC(OC)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4160 to 1.4190 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only