N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 mL
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 115690010
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
78.70 
Save 12.05 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
78.70 
Save 12.05 (15%)
Add to cart
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
Catalog number115690010
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
78.70 
Save 12.05 (15%)
-
Add to cart
Chemical Identifiers
IUPAC NameN,N-dimethylacetamide
Molecular FormulaC4H9NO
InChI KeyFXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILESCN(C)C(C)=O
Molecular Weight (g/mol)87.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
GC>=98.5 %
Water=<0.3 %
Refractive index1.4370 to 1.4390 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only