N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
5 mL
1 L
2.5 L
Packaging:
View all
Metal Drum
Catalog number 115690250
Price (USD)/ Each
889.65
Online exclusive
1047.00 
Save 157.35 (15%)
-
Add to cart
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Price (USD)/ Each
889.65
Online exclusive
1047.00 
Save 157.35 (15%)
Add to cart
N,N-Dimethylacetamide, 99%, pure, Thermo Scientific Chemicals
Catalog number115690250
Price (USD)/ Each
889.65
Online exclusive
1047.00 
Save 157.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC NameN,N-dimethylacetamide
Molecular FormulaC4H9NO
InChI KeyFXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILESCN(C)C(C)=O
Molecular Weight (g/mol)87.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
GC>=98.5 %
Water=<0.3 %
Refractive index1.4370 to 1.4390 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only