Thermo Scientific Chemicals

Dimethyl 1,3-acetonedicarboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 115705000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Dimethyl 1,3-acetonedicarboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 115705000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 115705000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1830-54-2
IUPAC Name
1,5-dimethyl 3-oxopentanedioate
Molecular Formula
C7H10O5
InChI Key
RNJOKCPFLQMDEC-UHFFFAOYSA-N
SMILES
COC(=O)CC(=O)CC(=O)OC
Refractive index
1.4430 to 1.4450 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text