Dimethyl acetylenedicarboxylate, 98%, Thermo Scientific Chemicals
Dimethyl acetylenedicarboxylate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl acetylenedicarboxylate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
500 g
Catalog number 115710250
Price (USD)/ Each
79.65
Online exclusive
88.50 
Save 8.85 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
79.65
Online exclusive
88.50 
Save 8.85 (10%)
Add to cart
Dimethyl acetylenedicarboxylate, 98%, Thermo Scientific Chemicals
Catalog number115710250
Price (USD)/ Each
79.65
Online exclusive
88.50 
Save 8.85 (10%)
-
Add to cart
Chemical Identifiers
CAS762-42-5
IUPAC Name1,4-dimethyl but-2-ynedioate
Molecular FormulaC6H6O4
InChI KeyVHILMKFSCRWWIJ-UHFFFAOYSA-N
SMILESCOC(=O)C#CC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4460 to 1.4500 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only