Thermo Scientific Chemicals

3-Dimethylaminobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 115751000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

3-Dimethylaminobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 115751000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 115751000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-64-9
IUPAC Name
3-(dimethylamino)benzoic acid
Molecular Formula
C9H11NO2
InChI Key
NEGFNJRAUMCZMY-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=CC(=C1)C(O)=O
Appearance (Color)
Light yellow to beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
150°C to 153°C
Titration with NaOH
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text