Thermo Scientific Chemicals

4-(Dimethylamino)benzonitrile, 95%, Thermo Scientific Chemicals

Catalog number: 115780250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(Dimethylamino)benzonitrile, 95%, Thermo Scientific Chemicals

Catalog number: 115780250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 115780250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1197-19-9
IUPAC Name
4-(dimethylamino)benzonitrile
Molecular Formula
C9H10N2
InChI Key
JYMNQRQQBJIMCV-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(C=C1)C#N
Appearance (Form)
Crystalline powder
Melting point
70°C to 76°C
Titration with HClO4
>=94 %
Appearance (Color)
Light brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text