Thermo Scientific Chemicals

4-Dimethylaminocinnamaldehyde, 98%

Catalog number: 115790250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Dimethylaminocinnamaldehyde, 98%

Catalog number: 115790250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 115790250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6203-18-5
IUPAC Name
(2E)-3-[4-(dimethylamino)phenyl]prop-2-enal
Molecular Formula
C11H13NO
InChI Key
RUKJCCIJLIMGEP-ONEGZZNKSA-N
SMILES
CN(C)C1=CC=C(\C=C\C=O)C=C1
Appearance (Form)
Powder
Titration with HClO4
>=97.5 %
Appearance (Color)
Yellow to orange
Melting point
137°C to 142°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text