Thermo Scientific Chemicals

2-Dimethylaminoethyl chloride hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 115820025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

2-Dimethylaminoethyl chloride hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 115820025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 115820025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4584-46-7
IUPAC Name
hydrogen (2-chloroethyl)dimethylamine chloride
Molecular Formula
C4H11Cl2N
InChI Key
LQLJZSJKRYTKTP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CN(C)CCCl
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Chloride (Cl)
24.1 to 25.1 % (ionic chloride)
Appearance (Color)
White to brown-yellow
Melting point
203°C to 210°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text